Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCA2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523708

Catalog No. PIPA523708


Add to cart

Description

Description

This antibody is predicted to react with bovine based on sequence homology.

This gene encodes the human homologue of the mouse p(pink-eyed dilution) gene. The encoded protein is believed to be anintegral membrane protein involved in small molecule transport,specifically tyrosine - a precursor of melanin. Mutations in thisgene result in type 2 oculocutaneous albinism.
Specifications

Specifications

OCA2
Polyclonal
Unconjugated
OCA2
Liquid
OCA2
KLH conjugated synthetic peptide between 111-138 amino acids from the N-terminal region of human OCA2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q04671
BEY, BEY1, BEY2, BOCA, D15S12, EYCL, EYCL2, EYCL3, HCL3, P, PED, SHEP1, P protein, eye color 2 (central brown), eye color 3 (brown), hair color 3 (brown), melanocyte-specific transporter protein, oculocutaneous albinism II (pink-eye dilution homolog, mouse), pink-eyed dilution protein homolog, total brown iris pigmentation
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4948
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit