Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCC1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520672

Catalog No. PIPA520672


Add to cart

Description

Description

Despite the predicted molecular weight, OCC-1 from cell lines often migrates at ~28kDa in SDS-PAGE. A suggested positive control is 293 cell lysate. PA5-20672 can be used with blocking peptide PEP-0790.

OCC-1 (Overexpressed in colon carcinoma-1) was initially identified as a novel human cDNA overexpressed in a colon carcinoma cell line. Elevated levels of OCC-1 mRNA were observed in three of eight colon carcinomas, suggesting that OCC-1 may be a hallmark of only a subset of colon carcinomas. It is highly expressed in skeletal muscle, kidney, placenta, and pancreas tissues, with low to no expression in heart, brain, lung and liver tissues. At least two different OCC-1 mRNA species are known to exist.
Specifications

Specifications

OCC1
Polyclonal
Unconjugated
C12orf75
Liquid
C12orf75
a 7 amino acid peptide near the carboxy terminus of human OCC-1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8TAD7
AGD3, OCC-1, OCC1; overexpressed in colon carcinoma 1 protein; overexpressed in colon carcinoma-1
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
387882
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit