Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCIAD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA520835
Description
A suggested positive control is A549 cell lysate. PA5-20835 can be used with blocking peptide PEP-0949.
OCIAD2 Polyclonal antibody specifically detects OCIAD2 in Human samples. It is validated for Western Blot.Specifications
OCIAD2 | |
Polyclonal | |
Unconjugated | |
OCIAD2 | |
1810027I20Rik; OCIA domain containing 2; OCIA domain-containing protein 2; OCIAD2; Ovarian carcinoma immunoreactive antigen-like protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
132299 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q56VL3 | |
OCIAD2 | |
An 18 amino acid peptide from near the carboxy terminus of human OCIAD2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction