Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OCRL/INPP5b Mouse anti-Human, Rat, Clone: N166A/26, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABN609
Description
Detect Inositol polyphosphate 5-phosphatase OCRL-1 using this mouse monoclonal antibody, Anti-OCRL/INPP5b Antibody, clone N166A/26 validated for use in western blotting and immunohistochemistry.
Specifications
OCRL/INPP5b | |
Monoclonal | |
Unconjugated | |
NP_000267 | |
Mouse | |
Protein G purified | |
Primary | |
2-8°C for one year | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
N166A/26 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
OCRL, INPP5F, OCRL1 | |
Recombinant protein corresponding to human OCRL/INPP5b. | |
100 μg | |
Human, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction