Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oct-2 Rat, PE, Clone: 9A2, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 562838

Catalog No. BDB562838


Add to cart

Description

Description

The 9A2 monoclonal antibody specifically binds to Oct-2 that has a molecular weight of approximately 50-60 kDa. The 9A2 antibody was generated against the 44 N-terminal amino acids common to mouse Oct-2 isoforms and crossreacts with both human and mouse Oct-2. Oct-2 is a POU class 2 homeobox 2 transcription factors (POU2F2) that is also known as OCT2, OTF2, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2 and Octamer-binding transcription factor 2. The POU domain in the Oct-2 protein is required for low and high affinity cooperative binding of the octomer sequence and heptamer site in immunoglobulin promoters and promoters for other genes. Oct-2 is expressed in B cells, activated CD4+ and CD8+ T cells, monocytes/macrophages and subsets of dendritic cells. Oct-2 is reportedly associated with neoplasms including Diffuse Large B-cell lymphoma and Nodular Lymphocyte predominant Hodgkin's lymphoma. Studies with knockout mouse models suggest Oct-2 is required for B cell maturation, but not for B cell commitment, and for B cell production of IL-6.

Host Species: Rat
Clone: 9A2
Isotype: IgG2a, κ
Species Reactivity: Hu, Ms
Immunogen: Mouse Oct-2 amino acids 1-44 Recombinant Protein

Flow Cytometry, Intracellular Staining

Specifications

Specifications

OCT2
9A2
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Human, Murine
Flow Cytometry
0.2mg/mL
Oct2, OTF-2, OCT2, OTF2, Oct2, NF-A2, POU2F2, Pou2f2, Otf2
Affinity Purified
Mouse Oct-2 amino acids 1-44 Recombinant Protein
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit