Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OCT1 (POU2F1) Rabbit anti-Bovine, Equine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523374

Catalog No. PIPA523374


Add to cart

Description

Description

OCT1/OTF1 is a protein belonging to the POU domain of transcription factors. OCT1, Octamer binding transcription factor 1, binds to an octamer motif (5 ft.-ATTTGCAT-3 ft.) and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes for immunoglobulins. POU domain proteins contain a bipartite DNA-binding domain, which is divided by a flexible linker that enables them to adopt various monomer configurations on DNA. The versatility of POU protein operation is conferred at the dimerization level. POU dimer from the OCT1 gene formed on the palindromic OCT factor recognition element, or PORE, could recruit the transcriptional coactivator OBF1. This octamer sequence within immunoglobulin promoters are sufficient for lymphoid-specific promoter activity in lymphoid cells. OCT1 regulate the expansion and/or maintece of mature B cells. In its absence mature B-cell pull is strongly reduced.
Specifications

Specifications

OCT1 (POU2F1)
Polyclonal
Unconjugated
POU2F1
Liquid
POU2F1
Synthetic peptide (428 CNRRQKEKRINPPSSGGTS 446) of OCT1/OTF1 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Equine, Human, Mouse, Rat
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
P14859, P25425, P31503
OCT1, OTF1, oct-1B, NF-A1, OTF-1, Octamer-binding transcription factor-1, POU domain, class 2, transcription factor 1, oct-1, octamer-binding protein 1, octamer-binding transcription factor 1
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100051363, 171068, 18986, 512840, 5451
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit