Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF2 (p Ser796) Rabbit anti-Human, Chimpanzee, Rhesus Macaque, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11085525

Catalog No. NB11085525


Add to cart

Description

Description

ODF2 Polyclonal specifically detects ODF2 in Human, Chimpanzee, Rhesus Macaque samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

ODF2 (p Ser796)
1 mg/ml
Western Blot 1:200-1:2000, ELISA 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Q5BJF6
Cenexin, FLJ44866, MGC111096, MGC9034, ODF2/1, ODF2/2, ODF8484-kD, outer dense fiber of sperm tails 2, outer dense fiber protein 2, sperm tail structural protein
Rabbit
IgG
Immunogen affinity purified
RUO
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody is specific for human Cenexin-1 protein phosphorylated at [p Ser796].
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
ODF2
This affinity purified ODF2 [p Ser796] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to residues near S796 of human ODF2. (Uniprot: Q5BJF6)
95.4 kDa
0.1 mg
Phospho Specific
Primary
4957
Human, Chimpanzee, Rhesus Macaque
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only