Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ODF3B Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524301

Catalog No. PIPA524301

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

ODF3B
Polyclonal
PBS with 0.09% sodium azide
A8MYP8, Q5M8M2
ODF3L3, orf2 5′ to PD-ECGF/TP, outer dense fiber protein 3-like protein 3, outer dense fiber protein 3B
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
440836, 70113
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ODF3B
Liquid
ODF3B
KLH conjugated synthetic peptide between 19-47 amino acids from the N-terminal region of human ODF3B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit