Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-OGDH, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219622
Description
OGDH Polyclonal specifically detects OGDH in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
OGDH | |
Polyclonal | |
Unconjugated | |
PBS, 1% BSA, 20% Glycerol (pH7) with 0.025% Proclin 300 | |
2-oxoglutarate dehydrogenase complex component E1, 2-oxoglutarate dehydrogenase, mitochondrial, AKGDH, alpha KGD, alpha-KGD, E1k, OGDC, oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide), oxoglutarate decarboxylase, oxoglutarate dehydrogenase (succinyl-transferring) | |
Rabbit | |
116 kDa | |
0.1 mL | |
Primary | |
Species reactivity based on sequence identity: Chicken (87%), Xenopus laevis (84%), Zebrafish (81%), Xenopus tropicalis (83%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0.65mg/mL | |
Western Blot 1:500-1:10000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
Q02218 | |
OGDH | |
Recombinant protein encompassing a sequence within the C-terminus region of human OGDH. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
4967 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only