Learn More
OMP Rabbit anti-Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00020G
Catalog No. PIOSR00020G
Add to cart
Description
Description
The peptide is homologous with the corresponding sequence derived from OMP protein in mouse. Reconstitute in 500 uL of sterile water.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
Specifications
Specifications
OMP
Polyclonal
PBS with no preservative
Q64288, P08523
Olfactory neuronal-specific protein
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18378, 24612
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Omp
Lyophilized
OMP
Synthetic peptide from rat Olfactory Marker Protein.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse, Rat