Learn More
OMP Sheep anti-Mouse, Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Supplier:  Thermo Scientific OSS00002G
Catalog No. PIOSS00002G
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
The peptide is homologous with the corresponding sequence derived from OMP protein in mouse.
Olfactory marker protein (OMP) is an abundant, 19-kDa, cytosolic protein that is almost exclusively expressed in mature, functioning, olfactory neurons but not in the neural precursor basal cells. The OMP gene structure and protein sequence are highly conserved between mouse, rat and human. Its tissue-specific expression in the receptor cells together with the results of the mouse knockout studies show that OMP represents a novel modulatory component of the odor detection/signal transduction cascade. May act as a modulator of the olfactory signal-transduction cascade. Interacts with BEX1 and BEX2. Uniquely associated with mature olfactory receptor neurons. Belongs to the olfactory marker protein family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OMP
Polyclonal
Omp
Olfactory neuronal-specific protein
Sheep
Ammonium sulfate precipitation
RUO
18378, 24612
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Q64288, P08523
OMP
Synthetic peptide from rat Olfactory Marker Protein.
500 μg
Primary
Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions