Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LS189744 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA132773
Description
PA1-32773 detects Olfactory receptor LS189744 from human samples.
LS189744 Polyclonal antibody specifically detects LS189744 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
LS189744 | |
Polyclonal | |
Unconjugated | |
OR6K3 | |
OR6K3 | |
Synthetic 18 amino acid peptide from C-terminus of human OR6K3 | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q8NGY3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
391114 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction