Learn More
Olfr15 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00030G
Catalog No. PIOSR00030G
Add to cart
Description
Description
Reconstitute with 500 µL of distilled water. Centrifuge to remove any insoluble material.
Belongs to the G-protein coupled receptor 1 family.
Specifications
Specifications
Olfr15
Polyclonal
PBS with no preservative
P23275
Olfactory receptor Olfr15, Odorant receptor OR3. GENE NAME: Olfr15.
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18312
Immunohistochemistry, Western Blot
Unconjugated
Olfr15
Lyophilized
OLFR15
A synthetic peptide from n-terminal region of mouse olfactory receptor family 256, subfamily 17
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse