Learn More
Olfr154 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00039W
Catalog No. PIOSR00039W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material. OSR00039W can be used with blocking peptide GSM00009P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
Olfr154
Polyclonal
whole serum with no preservative
Q9QY00
Olfactory receptor 154, Olfactory receptor 175-1, ORL782, Or912-93. GENE NAME: Name: Olfr154, Mor175-1, Olfr4-3, Or912-93, Or93
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Olfr154
Lyophilized
OLFR154
Synthetic peptide from mouse olfactory receptor family 175, subfamily 1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
27216