Learn More
Olfr412 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00211W
Catalog No. PIOSR00211W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome.
Specifications
Specifications
Olfr412
Polyclonal
whole serum with no preservative
Lyophilized
OLFR412
A synthetic peptide from c-terminal region of mouse olfactory receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
258153
Immunohistochemistry
Unconjugated
Olfr412
Olfactory receptor Olfr412, olfactory receptor 412, Olfr412
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse