Learn More
Olfr49 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00040W
Catalog No. PIOSR00040W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material. OSR00040W can be used with blocking peptide GSM00007P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
Olfr49
Polyclonal
whole serum with no preservative
Q9Z1V0
ORL451, C6, Q8VFB7_MOUSE, Olfactory receptor MOR118-1, Olfactory receptor Olfr49. GENE NAME: Olfr49
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Olfr49
Lyophilized
OLFR49
Synthetic peptide from mouse olfactory receptor family 118, subfamily 1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18348