Learn More
Olfr56 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00014W
Catalog No. PIOSR00014W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. Reconstitute in 100 uL of sterile water. OSR00014W can be used with blocking peptide GSM00014P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals.
Specifications
Specifications
Olfr56
Polyclonal
whole serum with no preservative
Q8VGD6
Olfactory receptor Olfr56, ORL436, G7. GENE NAME: Olfr56
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Olfr56
Lyophilized
OLFR56
Synthetic peptide from mouse olfactory receptor family 276, subfamily 1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
18356