Learn More
Olfr642 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00027G
Catalog No. PIOSR00027G
Add to cart
Description
Description
Reconstitute with 500 µL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
Olfr642
Polyclonal
PBS with no preservative
Lyophilized
OLFR642
Synthetic peptide from mouse olfactory receptor family 13, subfamily 6, 2 and 5.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Olfr642
Olfactory receptor S85, Olfactory receptor 642, Olfactory receptor Olfr642, Olfactory receptor MOR13-6, S85, ORL473, Olfr642, MOR13-2, Olfr, MOR13-6, MOR13-2, MOR13-5
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
258326