Learn More
Olfr653 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00033G
Catalog No. PIOSR00033G
Add to cart
Description
Description
Reconstitute with 500 µL of distilled water. Centrifuge to remove any insoluble material.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
Olfr653
Polyclonal
PBS with no preservative
Lyophilized
OLFR653
Synthetic peptide from mouse olfactory receptor family 33, subfamily 1.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry, Western Blot
Unconjugated
Olfr653
Olfactory receptor MOR33-1, Olfactory receptor 653, Olfactory receptor Olfr653, ORL465, S19
Rabbit
IgG
500 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
57250