Learn More
OLFR749 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00007W
Catalog No. OSR00007W
Add to cart
Description
Description
Glycerol (1:1) may be added for additional stability. OSR00007W can be used with blocking peptide GSO00001P.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OLFR749
Polyclonal
whole serum with no preservative
Lyophilized
Olfr749
Synthetic peptide from mouse olfactory receptor family 106, subfamily 1.
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
56858
Immunohistochemistry, Western Blot
Unconjugated
Olfr749
Odorant receptor S1, Olfactory receptor MOR106-1, ORL461. GENE NAME: Olfr749
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse