Learn More
OR11H4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSR00023G
Catalog No. PIOSR00023G
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 500 μL of distilled water. Centrifuge to remove any insoluble material.
Putative odorant receptor. Multi-pass membrane protein. Belongs to the G-protein coupled receptor 1 family.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OR11H4
Polyclonal
OR11H4
Olfactory receptor OR14-36. GENE NAME: OR11H4
Rabbit
Ammonium sulfate precipitation
RUO
390442
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry
Unconjugated
Q8NGC9
OR11H4
A synthetic peptide from c-terminal region of human olfactory receptor
500 μg
Primary
Human
Antibody
IgG
Recommendations
Recommendations