Learn More
OR11H4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00008W
Catalog No. PIOSR00008W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water. Centrifuge to remove any insoluble material.
Putative odorant receptor. Multi-pass membrane protein. Belongs to the G-protein coupled receptor 1 family.
Specifications
Specifications
OR11H4
Polyclonal
whole serum with no preservative
Q8NGC9
Olfactory receptor OR14-36. GENE NAME: OR11H4
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry
Unconjugated
OR11H4
Lyophilized
OR11H4
A synthetic peptide from c-terminal region of human olfactory receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
390442