Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLFM1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512290

Catalog No. PIPA512290

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with chicken, mouse and rat based on sequence homology.

Olfm1 shares extensive sequence similarity with the rat neuronal olfactomedin-related ER localized protein. While the exact function of this protein is not known, its abundant expression in brain suggests that it may have an essential role in nerve tissue.
Specifications

Specifications

Olfm1
Polyclonal
PBS with 0.09% sodium azide
Q99784
Noelin; Neuronal olfactomedin-related ER localized protein; Olfactomedin-1; OLFM1; NOE1; NOEL1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10439
Immunohistochemistry, Western Blot
Unconjugated
OLFM1
Liquid
OLFM1
KLH conjugated synthetic peptide between 71-100 amino acids from the N-terminal region of human Olfm1
Protein G
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit