Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLFML2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525706

Catalog No. PIPA525706

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

OLFML2A contains one olfactomedin-like domain. Its exact function is unknown. There are three named isoforms produced by alternative splicing.
Specifications

Specifications

OLFML2A
Polyclonal
PBS with 0.09% sodium azide
Q68BL7
PRO34319, olfactomedin-like protein 2A, photomedin-1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
169611
Immunohistochemistry, Western Blot
Unconjugated
OLFML2A
Liquid
OLFML2A
KLH conjugated synthetic peptide between 6-34 amino acids from the N-terminal region of human OLFML2A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit