Learn More
anti-Olig2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner 29310002
Catalog No. 29-310-002
Add to cart
Description
Description
Olig2 Polyclonal antibody specifically detects Olig2 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.