Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OLIG3 Mouse anti-Human, Mouse, Clone: 257934, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523956

Catalog No. PIMA523956


Add to cart

Description

Description

In Western blots, no cross-reactivity with recombit human (rh) Olig1 or rhOlig2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

Olig3
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q7RTU3, Q6PFG8
BHLHB7, BHLHB7, Bhlhb7, bHLHb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
167826, 94222
Immunohistochemistry, Western Blot
257934
Unconjugated
OLIG3
Lyophilized
OLIG3
E. coli-derived recombit human Olig3 Met1-Lys272
Protein A/G
RUO
Antibody
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit