Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-OPA1, HRP, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11055290H

Catalog No. NB11055290H


Add to cart

Description

Description

OPA1 Polyclonal specifically detects OPA1 in Human, Mouse, Rat, Porcine, Chicken, Zebrafish samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications

Specifications

OPA1
HRP
PBS with No Preservative
Affinity Purified
OPA1
A synthetic peptide made to an internal region within residues 500-600 of human OPA1. [Swiss-Prot: O60313]
111 kDa
0.1 ml
Apoptosis, Mitochondrial Fusion Proteins, Neurodegeneration, Neuroscience, Sensory Systems, Vision
Primary
4976
Human, Mouse, Rat, Porcine, Chicken
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot
Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting
O60313
FLJ12460, KIAA0567dynamin-like 120 kDa protein, mitochondrial, MGM1, mitochondrial dynamin-like GTPase, NPGlargeG, NTGmitochondrial dynamin-like 120 kDa protein, optic atrophy 1 (autosomal dominant), Optic atrophy protein 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Human, mouse, rat and chicken. Predicted to react with primate based on 100% sequence homology. Immunogen sequence has 94% homology to Zebrafish.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only