Learn More
OPRM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA3203
Catalog No. PA3203
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA3-204 detects Nociceptin in human, rat and mouse samples. PA3-204 has successfully been used in immuncytochemical and immunohistochemical (paraffin) procedures. The PA3-204 immunogen is a synthetic peptide corresponding to the amino acids FGGFTGARKSAR
Specifications
Specifications
OPRM1
Polyclonal
whole serum with 0.02% sodium azide
P35372, P42866, P33535
mu-opioid receptor; MOR; MOP; MOR1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
OPRM1
Liquid
OPRM1
Synthetic peptide corresponding to the C-terminal amino acids L(387)ENLEAETAPLP(398) of OPRM1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
18390, 25601, 4988