Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma-1 Receptor Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512326

Catalog No. PIPA512326


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

OPRS1 is a receptor protein that interacts with a variety of psychotomimetic drugs, including cocaine and amphetamines. The receptor is believed to play an important role in the cellular functions of various tissues associated with the endocrine, immune, and nervous systems.
Specifications

Specifications

Sigma-1 Receptor
Polyclonal
Unconjugated
SIGMAR1
Liquid
SIGMAR1
KLH conjugated synthetic peptide between 47-81 amino acids from the N-terminal region of human OPRS1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Flow Cytometry, Western Blot
2.0 mg/mL
PBS with 0.09% sodium azide
Q99720
Sigma 1-type opioid receptor; Sigma1-receptor; Sigma1R; SIG-1R; hSigmaR1; SR31747-binding protein; SR-BP; Aging-associated gene 8 protein; OPRS1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10280
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit