Learn More
Opsin 5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00218W
Catalog No. PIOSR00218W
Add to cart
Description
Description
This product ships at ambient temperature. Reconstitute in 100 µL of distilled water.
Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
Specifications
Specifications
Opsin 5
Polyclonal
whole serum with no preservative
Q6U736, Q6VZZ7
Opsin-5, neuropsin, G-protein coupled receptor 136, G-protein coupled receptor PGR12, OPN5, opsin 5, GPR136, TMEM13
Rabbit
IgG
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunohistochemistry, Western Blot
Unconjugated
OPN5
Lyophilized
OPN5
A synthetic peptide from mouse opsin-5
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
221391, 316259, 353344