Learn More
OR10R2 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova
Rabbit polyclonal antibody raised against synthetic peptide of OR10R2.
Manufacturer: Abnova Corporation PAB26497
Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms. [provided by RefSeq
Specifications
OR10R2 | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (21 ug/mL) The optimal working dilution should be determined by the end user. | |
OR10R2 | |
OR1-8|OR10R2Q | |
Rabbit | |
Immunoaffinity chromatography | |
RUO | |
Primary | |
343406 | |
Human, Monkey |
Immunohistochemistry (PFA fixed) | |
Rabbit polyclonal antibody raised against synthetic peptide of OR10R2. | |
In PBS (0.09% sodium azide) | |
Liquid | |
OR10R2 | |
A synthetic peptide corresponding to 17 amino acids at C-terminus of human OR10R2. | |
50 ug | |
Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
|
Polyclonal | |
BLAST analysis of the peptide immunogen showed no homology with other human proteins. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only