Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Olfr417 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524134

Catalog No. PIPA524134


Add to cart

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications

Specifications

Olfr417
Polyclonal
PBS with 0.09% sodium azide
Q8NGY0
OR1-13, OR1-14, OR10X1P, olfactory receptor 10X1, olfactory receptor OR1-13 pseudogene, olfactory receptor OR1-14, olfactory receptor, family 10, subfamily X, member 1 pseudogene
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
128367, 258238
Western Blot
Unconjugated
OR10X1
Liquid
OLFR417, OR10X1
KLH conjugated synthetic peptide between 83-112 amino acids from the central region of human OR10X1
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit