Learn More
anti-OR1L3, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232321
Catalog No. NBP232321
Add to cart
Description
Description
OR1L3 Polyclonal antibody specifically detects OR1L3 in Human samples. It is validated for Western Blot.
Specifications
Specifications
OR1L3
0.5 mg/mL
Western Blot 1:1000
Q8NH93
olfactory receptor 1L3, Olfactory receptor OR9-28, olfactory receptor, family 1, subfamily L, member 3, OR9-28, OR9-DOlfactory receptor 9-D
Rabbit
IgG
Protein A purified
RUO
Primary
26735
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
OR1L3
This OR1L3 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 290-320 amino acids from the C-terminal region of human OR1L3.
36.633 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human