Learn More
OR1L8 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00258W
Catalog No. PIOSR00258W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OR1L8
Polyclonal
whole serum with no preservative
Q8NGR8
Olfactory receptor, family 1, subfamily L, member 8; OR1L8, OR9-24, HsOR9.6.6
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
OR1L8
Lyophilized
OR1L8
A synthetic peptide from human olfactory receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
138881