Learn More
OR2D3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00277W
Catalog No. PIOSR00277W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OR2D3
Polyclonal
whole serum with no preservative
Q8NGH3
OR2D3, OR11-89, Olfactory receptor 2D3, olfactory receptor, family 2, subfamily D, member 3
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
OR2D3
Lyophilized
OR2D3
A synthetic peptide from human olfactory receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
120775