Learn More
OR2V1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00287W
Catalog No. PIOSR00287W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OR2V1
Polyclonal
whole serum with no preservative
Q8NHB1
Olfactory receptor, family 2, subfamily V, member 1; OST265, Olfactory receptor 2V1
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
OR2V1
Lyophilized
OR2V1
A synthetic peptide from human olfactory receptor OR2V1
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
26693