Learn More
OR4A5 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen OSR00291W
Catalog No. PIOSR00291W
Add to cart
Description
Description
Reconstitute with 100 µL of distilled water.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OR4A5
Polyclonal
whole serum with no preservative
Q8NH83
Olfactory receptor, family 4, subfamily A, member 5; OR11-111, Olfactory receptor 4A5, Olfactory receptor OR11-111
Rabbit
IgG
RUO
Antibody
Polyclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
OR4A5
Lyophilized
OR4A5
A synthetic peptide from human olfactory receptor
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
81318