Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR8B8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524348

Catalog No. PIPA524348


Add to cart

Description

Description

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications

Specifications

OR8B8
Polyclonal
PBS with 0.09% sodium azide
Q15620
TPCR85, olfactory receptor 8B8, olfactory receptor OR11-316, olfactory receptor TPCR85, olfactory-like receptor JCG8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26493
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
OR8B8
Liquid
OR8B8
KLH conjugated synthetic peptide between 280-308 amino acids from the C-terminal region of human OR8B8
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit