Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-ORAOV1, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals 44910002

Catalog No. 44-910-002


Add to cart

Description

Description

ORAOV1 Polyclonal antibody specifically detects ORAOV1 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

ORAOV1
Unconjugated
20mM Potassium Phosphate (pH 7.0) and 0.15M NaCl with No Preservative
Affinity Purified
ORAOV1
This antibody is specific for the entire range of the target protein.
Immunogen affinity purified
RUO
Primary
220064
Human
ELISA, Western Blot
Western Blot 1:5000-1:20000, ELISA 1:100-1:2000
Q8WV07
oral cancer overexpressed 1, TAOS1oral cancer overexpressed protein 1-A, Tumor-amplified and overexpressed sequence 1
Rabbit
IgG
0.05 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This product is specific for Human ORAOV1.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only