Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Orc2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP133189

Catalog No. NBP133189


Add to cart

Description

Description

Orc2 Polyclonal antibody specifically detects Orc2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Orc2
0.45mg/mL
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000
Affinity Purified
ORC2
Recombinant protein encompassing a sequence within the center region of human ORC2. The exact sequence is proprietary.
66 kDa
0.1 ml
Cell Cycle and Replication
Primary
4999
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.01% Thimerosal
ORC2Lorigin recognition complex subunit 2, origin recognition complex protein 2 homolog, origin recognition complex, subunit 2, origin recognition complex, subunit 2 (yeast homolog)-like, origin recognition complex, subunit 2 homolog, origin recognition complex, subunit 2 homolog (yeast), origin recognition complex, subunit 2-like, origin recognition complex, subunit 2-like (yeast)
Rabbit
IgG
Immunogen affinity purified
RUO
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Based on sequence homology, it is expected to react with Mouse (84%) and Bovine (92%).
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only