Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528083

Catalog No. PIPA528083


Add to cart

Description

Description

PA5-28083 targets ORC3L in WB applications and shows reactivity with Human samples. The PA5-28083 immunogen is synthetic peptide corresponding to a region within amino acids 648 and 711 of Human ORC3L. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

The origin recognition complex (ORC) is a highly conserved six subunits protein complex essential for the initiation of the DNA replication in eukaryotic cells. Studies in yeast demonstrated that ORC binds specifically to origins of replication and serves as a platform for the assembly of additional initiation factors such as Cdc6 and Mcm proteins. The protein encoded by this gene is a subunit of the ORC complex. Studies of a similar gene in Drosophila suggested a possible role of this protein in neuronal proliferation and olfactory memory. Alternatively spliced transcript variants encoding distinct isoforms have been reported for this gene.
Specifications

Specifications

ORC3
Polyclonal
Unconjugated
ORC3
Liquid
ORC3
Synthetic peptide corresponding to a region within amino acids 648 and 711 of Human ORC3L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
Q9UBD5
LAT, LATHEO, ORC3L, homolog of latheo, Drosophila, origin recognition complex subunit 3, origin recognition complex subunit Latheo, origin recognition complex, subunit 3 honolog, origin recognition complex, subunit 3-like
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
23595
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit