Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ORC4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Supplier: Thermo Scientific PA517950
Description
This antibody is predicted to react with bovine, canine and rat based on sequence homology.
ORC4 Polyclonal antibody specifically detects ORC4 in Human samples. It is validated for Western Blot.Specifications
ORC4 | |
Polyclonal | |
Unconjugated | |
ORC4 | |
HSORC4; mMmORC4; Orc4; ORC4L; Orc4P; Origin recognition complex subunit 4; origin recognition complex, subunit 4; origin recognition complex, subunit 4 homolog; origin recognition complex, subunit 4-like | |
Goat | |
Antigen affinity chromatography | |
RUO | |
5000 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O43929 | |
ORC4 | |
Synthetic peptide sequence (DVRQWATSSLSWL) corresponding to the C-terminus amino acids of ORC4L | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction