Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Orexin R2/HCRTR2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems MAB52461
This item is not returnable.
View return policy
Description
Orexin R2/HCRTR2 Monoclonal specifically detects Orexin R2/HCRTR2 in Human samples. It is validated for Immunohistochemistry.Specifications
Orexin R2/HCRTR2 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
AAC39602 | |
HCRTR2 | |
NS0 mouse myeloma cell line transfected with human Orexin R2/HCRTR2 Met1-Trp444 Accession # AAC39602 | |
100 μg | |
Primary | |
Detects human Orexin R2/HCRTR2. In flow cytometry, stains human Orexin R2/HCRTR2 transfectants but not irrelevant transfectants. | |
Human | |
Purified |
Immunohistochemistry | |
456723 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
HCRTR2, hypocretin (orexin) receptor 2, Hypocretin receptor type 2, Orexin Receptor 2, orexin receptor type 2, OrexinR2, Ox-2-R, Ox2-R, OX2R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3062 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction