Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORMDL2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524302

Catalog No. PIPA524302


Add to cart

Description

Description

This antibody is predicted to react with bovine, rat and Xenopus and zebrafish based on sequence homology.

Negative regulator of sphingolipid synthesis.
Specifications

Specifications

ORMDL2
Polyclonal
PBS with 0.09% sodium azide
Q53FV1, Q9CQZ0
MST095, MSTP095, adoplin-2, ORM1-like protein 2, expressed in normal aorta
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
29095, 66844
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ORMDL2
Liquid
ORMDL2
KLH conjugated synthetic peptide between 53-81 amino acids from the central region of human ORML2
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit