Learn More
ODC1 Guinea Pig anti-Mouse, Polyclonal, Invitrogen™
Guinea Pig Polyclonal Antibody
Manufacturer:  Invitrogen PA136025
Catalog No. 01-672-215
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide. For long-term storage, aliquot and store at -20°C.
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
Specifications
Specifications
ODC1
Polyclonal
whole serum with no preservative
P00860
MGC103389, ODC
Guinea Pig
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry (Frozen)
Unconjugated
Odc1
Lyophilized
ODC1
Recombit human ODC produced in E. coli.
50 μL
4° C
Primary
18263