Learn More
ODC1 Rabbit anti-Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136134
Catalog No. 01-672-290
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 50-100 µL of distilled water, and dilute further in 0.1M PBS with 1% BSA and 0.1% sodium azide.
Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
Specifications
Specifications
ODC1
Polyclonal
whole serum with no preservative
P00860
MGC103389, ODC
Rabbit
IgG
RUO
Antibody
Polyclonal
Mouse
Immunohistochemistry (Frozen)
Unconjugated
Odc1
Lyophilized
ODC1
Recombit human ODC produced in E. coli.
50 μL
4° C
Primary
18263