Learn More
Orphanin FQ Rabbit anti-Non-human primate, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA129768
Catalog No. PIPA129768
Add to cart
Description
Description
PA1-29768 detects Orphanin FQ from non-human primate, rat samples. PA1-29768 has been successfully used in immunohistochemistry applications. The PA1-29768 immunogen is: FGGFTGARKSARKLANQ Corresponding to residues 122-138 of rat Orphanin FQ Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.
Specifications
Specifications
Orphanin FQ
Polyclonal
whole serum with no preservative
Q13519, Q62923
PNOC
FGGFTGARKSARKLANQ Corresponding to residues 122-138 of rat Orphanin FQ
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
25516, 5368
Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Unconjugated
PNOC
Liquid
Rabbit
IgG
RUO
Antibody
Polyclonal
Non-human Primate, Rat