Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OSMR Mouse anti-Human, Clone: 469221, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524190

Catalog No. PIMA524190

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody stains human ADRA1B transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

OSMR
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q99650
oncostatin M receptor, MGC150626, MGC150627, MGC75127
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
9180
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
469221
Unconjugated
OSMR
Lyophilized
OSMR
Mouse myeloma cell line NS0-derived recombit human OSM R beta Glu28-Ser739
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit