Learn More
anti-OSR1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP131198
Catalog No. NBP131198
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
OSR1 Polyclonal antibody specifically detects OSR1 in Human samples. It is validated for Western Blot.
Specifications
Specifications
OSR1
1.0mg/mL
Western Blot 1:500-1:3000
Affinity Purified
OSR1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human OSR1. The exact sequence is proprietary.
30 kDa
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Zebrafish (89%), Xenopus laevis (94%), and Xenopus tropicalis (94%). Not yet tested in any other species.
Western Blot
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
ODDodd-skipped (Drosophila) homolog, odd-skipped homolog, odd-skipped related 1 (Drosophila), protein odd-skipped-related 1
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
130497
Human