Learn More
OSR1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA528962
Catalog No. PIPA528962
Add to cart
Description
Description
PA5-28963 targets AKR1D1 in WB applications and shows reactivity with Human samples. The PA5-28963 immunogen is recombit fragment corresponding to a region within amino acids 1 and 326 of Human AKR1D1.
Specifications
Specifications
OSR1
Polyclonal
Unconjugated
OSR1
Liquid
OSR1
Synthetic peptide corresponding to a region within amino acids 107 and 204 of Human OSR1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8TAX0
ODD, odd-skipped homolog, protein odd-skipped-related 1
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
130497